Assay Details
| Gene Symbol: WDR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030628.1 | NP_001025799.1 | 4 - 5 | 826 | 63 | |
| XM_015294665.1 | XP_015150151.1 | 3 - 4 | 917 | 63 | ||
| XM_015294666.1 | XP_015150152.1 | 3 - 4 | 917 | 63 | ||
| GenBank mRNA | AJ719275.1 | - | 4 - 5 | 826 | 63 | |
More Information
Set Membership: |
|

WDR24
Genomic Map