Assay Details
| Gene Symbol: HNF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030855.1 | NP_001026026.1 | 6 - 7 | 731 | 61 | |
| XM_015296366.2 | XP_015151852.1 | 6 - 7 | 1116 | 61 | ||
| XM_025142195.1 | XP_024997963.1 | 6 - 7 | 756 | 61 | ||
| XM_025142196.1 | XP_024997964.1 | 6 - 7 | 753 | 61 | ||
| GenBank mRNA | AY700581.1 | - | 6 - 7 | 731 | 61 | |
More Information
Set Membership: |
|

HNF4A
Genomic Map