Assay Details
Gene Symbol: TPD52L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030858.1 | NP_001026029.1 | 4 - 5 | 398 | 66 | |
GenBank mRNA | AJ720891.1 | - | 4 - 5 | 398 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
