Assay Details
| Gene Symbol: MAPRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030860.1 | NP_001026031.1 | 5 - 6 | 616 | 67 | |
| XM_015296403.2 | XP_015151889.2 | 5 - 6 | 652 | 67 | ||
| GenBank mRNA | AJ719807.1 | - | 5 - 6 | 616 | 67 | |
| CR390770.1 | - | 3 - 4 | 425 | 67 | ||
More Information
Set Membership: |
|

MAPRE1
Genomic Map