Assay Details
| Gene Symbol: NOC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030876.1 | NP_001026047.1 | 17 - 18 | 2025 | 63 | |
| GenBank mRNA | AJ720345.1 | - | 17 - 18 | 2025 | 63 | |
More Information
Set Membership: |
|

NOC2L
Genomic Map