Assay Details
| Gene Symbol: RRAGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030896.1 | NP_001026067.1 | 1 - 2 | 295 | 74 | |
| GenBank mRNA | AJ719695.1 | - | 1 - 2 | 295 | 74 | |
More Information
Set Membership: |
|

RRAGC
Genomic Map