Assay Details
Gene Symbol: ACTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031063.1 | NP_001026234.1 | 4 - 5 | 812 | 68 | |
XM_015284056.2 | XP_015139542.2 | 5 - 6 | 885 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
