Assay Details
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031069.1 | NP_001026240.1 | 1 - 2 | 283 | 128 | |
XM_025148725.1 | XP_025004493.1 | 1 - 2 | 340 | 128 | ||
GenBank mRNA | AJ719888.1 | - | 1 - 2 | 283 | 128 | |
AY452479.1 | - | 1 - 2 | 264 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
