Assay Details
| Gene Symbol: ORAOV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031186.1 | NP_001026357.1 | 2 - 3 | 254 | 59 | |
| XM_015286805.2 | XP_015142291.1 | 2 - 3 | 239 | 59 | ||
| XM_015286807.2 | XP_015142293.1 | 2 - 3 | 239 | 59 | ||
| XM_025150699.1 | XP_025006467.1 | 2 - 3 | 239 | 59 | ||
| XM_025150700.1 | XP_025006468.1 | 2 - 3 | 239 | 59 | ||
| GenBank mRNA | AJ720231.1 | - | 2 - 3 | 254 | 59 | |
| CR387475.1 | - | 2 - 3 | 187 | 59 | ||
More Information
Set Membership: |
|

ORAOV1
Genomic Map