Assay Details
| Gene Symbol: SLC16A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031222.1 | NP_001026393.1 | 4 - 5 | 593 | 100 | |
| XM_015288131.2 | XP_015143617.2 | 5 - 6 | 684 | 100 | ||
| XM_015288132.2 | XP_015143618.2 | 4 - 5 | 479 | 100 | ||
| XM_015288133.2 | XP_015143619.2 | 5 - 6 | 659 | 100 | ||
| XM_015288134.2 | XP_015143620.2 | 5 - 6 | 875 | 100 | ||
| XM_015288135.2 | XP_015143621.2 | 4 - 5 | 757 | 100 | ||
| GenBank mRNA | AJ720344.1 | - | 4 - 5 | 593 | 100 | |
More Information
Set Membership: |
|

SLC16A9
Genomic Map