Assay Details
| Gene Symbol: PIP5K1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031422.1 | NP_001026593.1 | 6 - 7 | 752 | 81 | |
| XM_015280273.2 | XP_015135759.1 | 7 - 8 | 819 | 81 | ||
| XM_015280274.2 | XP_015135760.1 | 8 - 9 | 844 | 81 | ||
| XM_015280275.2 | XP_015135761.1 | 8 - 9 | 903 | 81 | ||
| XM_015280276.2 | XP_015135762.1 | 8 - 9 | 896 | 81 | ||
| XM_015280277.2 | XP_015135763.1 | 8 - 9 | 1027 | 81 | ||
| XM_015280278.2 | XP_015135764.1 | 7 - 8 | 778 | 81 | ||
| XM_015280279.1 | XP_015135765.1 | 7 - 8 | 687 | 81 | ||
| XM_015280280.2 | XP_015135766.1 | 7 - 8 | 795 | 81 | ||
| XM_015280281.2 | XP_015135767.1 | 6 - 7 | 635 | 81 | ||
| XM_015280282.2 | XP_015135768.1 | 7 - 8 | 680 | 81 | ||
| XM_015280283.2 | XP_015135769.1 | 7 - 8 | 819 | 81 | ||
| XM_015280285.2 | XP_015135771.1 | 8 - 9 | 842 | 81 | ||
| XM_015280286.2 | XP_015135772.1 | 7 - 8 | 801 | 81 | ||
| XM_025144712.1 | XP_025000480.1 | 9 - 10 | 928 | 81 | ||
| XM_025144713.1 | XP_025000481.1 | 8 - 9 | 968 | 81 | ||
| XM_025144714.1 | XP_025000482.1 | 8 - 9 | 869 | 81 | ||
| XM_025144715.1 | XP_025000483.1 | 8 - 9 | 770 | 81 | ||
| XM_025144716.1 | XP_025000484.1 | 6 - 7 | 659 | 81 | ||
| GenBank mRNA | AJ720576.1 | - | 6 - 7 | 752 | 81 | |
More Information
Set Membership: |
|

PIP5K1B
Genomic Map