Assay Details
| Gene Symbol: AvBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001194.1 | NP_001001194.1 | 2 - 3 | 133 | 95 | |
| GenBank mRNA | AY534893.1 | - | 1 - 2 | 55 | 95 | |
| AY621309.1 | - | 2 - 3 | 133 | 95 | ||
| DQ677638.1 | - | 1 - 2 | 55 | 95 | ||
| DQ858317.1 | - | 1 - 2 | 55 | 95 | ||
| DQ858330.1 | - | 1 - 2 | 55 | 95 | ||
| FJ882407.1 | - | 2 - 3 | 146 | 95 | ||
More Information
Set Membership: |
|

AvBD7
Genomic Map