Assay Details
| Gene Symbol: KRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001195.2 | NP_001001195.1 | 6 - 7 | 1330 | 61 | |
| GenBank mRNA | AB734684.1 | - | 6 - 7 | 1238 | 61 | |
| AB734685.1 | - | 6 - 7 | 1238 | 61 | ||
| AB734686.1 | - | 6 - 7 | 61 | |||
| AB734687.1 | - | 6 - 7 | 61 | |||
| AY574983.1 | - | 6 - 7 | 1330 | 61 | ||
More Information
Set Membership: |
|

KRT5
Genomic Map