Assay Details
| Gene Symbol: KRT75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001314.2 | NP_001001314.2 | 8 - 9 | 1459 | 75 | |
| GenBank mRNA | AB734677.1 | - | 8 - 9 | 1401 | 75 | |
| AB734678.1 | - | 8 - 9 | 1401 | 75 | ||
| AB734679.1 | - | 8 - 9 | 1401 | 75 | ||
| AY574984.1 | - | 8 - 9 | 1471 | 75 | ||
More Information
Set Membership: |
|

KRT75
Genomic Map