Assay Details
| Gene Symbol: VKORC1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001328.2 | NP_001001328.1 | 2 - 3 | 313 | 83 | |
| GenBank mRNA | BX930449.1 | - | 2 - 3 | 313 | 83 | |
More Information
Set Membership: |
|

VKORC1L1
Genomic Map