Assay Details
| Gene Symbol: PPARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001464.1 | NP_001001464.1 | 3 - 4 | 608 | 89 | |
| XM_015289929.2 | XP_015145415.1 | 4 - 5 | 490 | 89 | ||
| XM_015289937.2 | XP_015145423.1 | 4 - 5 | 580 | 89 | ||
| XM_015289940.2 | XP_015145426.1 | 4 - 5 | 1241 | 89 | ||
| XM_015289942.2 | XP_015145428.1 | 4 - 5 | 565 | 89 | ||
| XM_015289947.2 | XP_015145433.1 | 4 - 5 | 627 | 89 | ||
| XM_015289951.2 | XP_015145437.1 | 3 - 4 | 575 | 89 | ||
| XM_015289959.2 | XP_015145445.1 | 2 - 3 | 404 | 89 | ||
| XM_025150258.1 | XP_025006026.1 | 4 - 5 | 487 | 89 | ||
| GenBank mRNA | AF163809.1 | - | 3 - 4 | 608 | 89 | |
| AF470455.1 | - | 3 - 4 | 608 | 89 | ||
| BX931916.2 | - | 4 - 5 | 531 | 89 | ||
| EU049891.1 | - | 1 - 2 | 232 | 89 | ||
| KT318735.1 | - | 1 - 2 | 232 | 89 | ||
More Information
Set Membership: |
|

PPARA
Genomic Map