Assay Details
Gene Symbol: AvBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001608.2 | NP_001001608.2 | 1 - 2 | 288 | 84 | |
GenBank mRNA | AY534897.1 | - | 1 - 2 | 56 | 84 | |
AY621307.1 | - | 1 - 2 | 288 | 84 | ||
CR388932.1 | - | 1 - 2 | 238 | 84 | ||
DQ677636.1 | - | 1 - 2 | 84 | |||
DQ858302.1 | - | 1 - 2 | 128 | 84 | ||
DQ858315.1 | - | 1 - 2 | 128 | 84 | ||
DQ858342.1 | - | 1 - 2 | 128 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
