Assay Details
| Gene Symbol: AvBD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001609.2 | NP_001001609.1 | 2 - 3 | 265 | 142 | |
| GenBank mRNA | AY621312.2 | - | 2 - 3 | 249 | 142 | |
| BX274867.4 | - | 2 - 3 | 231 | 142 | ||
| CR388516.1 | - | 2 - 3 | 237 | 142 | ||
| DQ677641.1 | - | 2 - 3 | 215 | 142 | ||
| DQ858307.1 | - | 2 - 3 | 215 | 142 | ||
| DQ858320.1 | - | 2 - 3 | 215 | 142 | ||
| DQ858333.1 | - | 2 - 3 | 215 | 142 | ||
| DQ858347.1 | - | 2 - 3 | 215 | 142 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length greater than or equal to 101  3' Most  Recommended Assay  Gallus gallus  Probe spans exons | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 AvBD10
AvBD10 Genomic Map
Genomic Map Back To Top
 Back To Top