Assay Details
Gene Symbol: AvBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001610.2 | NP_001001610.1 | 3 - 4 | 508 | 72 | |
GenBank mRNA | AY621306.1 | - | 3 - 4 | 508 | 72 | |
DQ677635.1 | - | 2 - 3 | 228 | 72 | ||
DQ858301.1 | - | 2 - 3 | 228 | 72 | ||
DQ858314.1 | - | 2 - 3 | 228 | 72 | ||
DQ858327.1 | - | 2 - 3 | 228 | 72 | ||
DQ858341.1 | - | 2 - 3 | 228 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
