Assay Details
Gene Symbol: CYP3A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001751.2 | NP_001001751.1 | 2 - 3 | 215 | 64 | |
GenBank mRNA | AJ250337.1 | - | 2 - 3 | 216 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
