Assay Details
| Gene Symbol: CYP3A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001751.2 | NP_001001751.1 | 4 - 5 | 371 | 76 | |
| GenBank mRNA | AJ250337.1 | - | 4 - 5 | 372 | 76 | |
More Information
Set Membership: |
|

CYP3A5
Genomic Map