Assay Details
| Gene Symbol: LOC414835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001754.1 | NP_001001754.1 | 31 - 32 | 7899 | 67 | |
| XM_025147081.1 | XP_025002849.1 | 28 - 29 | 6543 | 67 | ||
| GenBank mRNA | AY312363.1 | - | 31 - 32 | 7899 | 67 | |
More Information
Set Membership: |
|

LOC414835
Genomic Map