Assay Details
| Gene Symbol: LOC414835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001754.1 | NP_001001754.1 | 3 - 4 | 1290 | 73 | |
| XM_015282913.2 | XP_015138399.1 | 3 - 4 | 6043 | 73 | ||
| XM_015282914.2 | XP_015138400.1 | 3 - 4 | 6043 | 73 | ||
| GenBank mRNA | AY312363.1 | - | 3 - 4 | 1290 | 73 | |
More Information
Set Membership: |
|

LOC414835
Genomic Map