Assay Details
| Gene Symbol: GSTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001776.1 | NP_001001776.1 | 3 - 4 | 411 | 156 | |
| XM_015284824.2 | XP_015140310.1 | 3 - 4 | 283 | 156 | ||
| XM_015284825.2 | XP_015140311.1 | 3 - 4 | 839 | 156 | ||
| GenBank mRNA | BX935545.1 | - | 2 - 3 | 148 | 156 | |
| L15387.2 | - | 3 - 4 | 411 | 156 | ||
More Information
Set Membership: |
|

GSTA2
Genomic Map