Assay Details
| Gene Symbol: GSTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001776.1 | NP_001001776.1 | 6 - 7 | 810 | 97 | |
| XM_015284824.2 | XP_015140310.1 | 6 - 7 | 682 | 97 | ||
| XM_015284825.2 | XP_015140311.1 | 6 - 7 | 1238 | 97 | ||
| GenBank mRNA | L15387.2 | - | 6 - 7 | 810 | 97 | |
More Information
Set Membership: |
|

GSTA2
Genomic Map