Assay Details
| Gene Symbol: AvBD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001780.1 | NP_001001780.1 | 1 - 2 | 58 | 91 | |
| GenBank mRNA | AY621315.1 | - | 1 - 2 | 58 | 91 | |
| AY701473.1 | - | 1 - 2 | 58 | 91 | ||
| DQ677644.1 | - | 1 - 2 | 58 | 91 | ||
| DQ858310.1 | - | 1 - 2 | 58 | 91 | ||
| DQ858323.1 | - | 1 - 2 | 58 | 91 | ||
| DQ858336.1 | - | 1 - 2 | 58 | 91 | ||
| DQ858350.1 | - | 1 - 2 | 58 | 91 | ||
More Information
Set Membership: |
|

AvBD13
Genomic Map