Assay Details
Gene Symbol: AvBD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001780.1 | NP_001001780.1 | 2 - 3 | 182 | 82 | |
GenBank mRNA | AY621315.1 | - | 2 - 3 | 182 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
