Assay Details
| Gene Symbol: CYP17A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001901.2 | NP_001001901.1 | 2 - 3 | 467 | 90 | |
| GenBank mRNA | M21406.1 | - | 2 - 3 | 471 | 90 | |
More Information
Set Membership: |
|

CYP17A1
Genomic Map