Assay Details
| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004399.1 | NP_001004399.1 | 2 - 3 | 219 | 71 | |
| GenBank mRNA | X60450.1 | - | 219 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LY86| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004399.1 | NP_001004399.1 | 2 - 3 | 219 | 71 | |
| GenBank mRNA | X60450.1 | - | 219 | 71 | ||
Set Membership: |
|