Assay Details
| Gene Symbol: AVEN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005791.1 | NP_001005791.1 | 1 - 2 | 291 | 78 | |
| GenBank mRNA | AJ583692.2 | - | 1 - 2 | 288 | 78 | |
| AJ719832.1 | - | 1 - 2 | 291 | 78 | ||
More Information
Set Membership: |
|

AVEN
Genomic Map