Assay Details
Gene Symbol: ACO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204188.2 | NP_989519.1 | 17 - 18 | 2624 | 59 | |
GenBank mRNA | AF428141.1 | - | 17 - 18 | 2624 | 59 | |
AJ719273.1 | - | 17 - 18 | 2243 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
