Assay Details
| Gene Symbol: SLC25A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204231.2 | NP_989562.2 | 3 - 4 | 790 | 67 | |
| GenBank mRNA | AB088686.1 | - | 3 - 4 | 747 | 67 | |
| AJ719767.1 | - | 3 - 4 | 790 | 67 | ||
More Information
Set Membership: |
|

SLC25A6
Genomic Map