Assay Details
Gene Symbol: NR0B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204593.1 | NP_989924.1 | 1 - 2 | 599 | 94 | |
XM_015302294.2 | XP_015157780.1 | 1 - 2 | 845 | 94 | ||
GenBank mRNA | AF202991.1 | - | 1 - 2 | 599 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
