Assay Details
| Gene Symbol: CD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204649.1 | NP_989980.1 | 7 - 8 | 1400 | 141 | |
| XM_015293164.2 | XP_015148650.1 | 7 - 8 | 1437 | 141 | ||
| GenBank mRNA | AY528652.1 | - | 6 - 7 | 1258 | 141 | |
| AY560014.1 | - | 6 - 7 | 1258 | 141 | ||
| AY560015.1 | - | 6 - 7 | 1258 | 141 | ||
| AY560016.1 | - | 6 - 7 | 1258 | 141 | ||
| DQ202315.1 | - | 6 - 7 | 1264 | 141 | ||
| Y12012.1 | - | 1400 | 141 | |||
More Information
Set Membership: |
|

CD4
Genomic Map