Assay Details
| Gene Symbol: APOA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204938.2 | NP_990269.1 | 2 - 3 | 210 | 83 | |
| GenBank mRNA | CR406924.1 | - | 2 - 3 | 197 | 83 | |
| Y16534.1 | - | 2 - 3 | 210 | 83 | ||
More Information
Set Membership: |
|

APOA4
Genomic Map