Assay Details
Gene Symbol: TBXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204940.2 | NP_990271.2 | 2 - 3 | 536 | 70 | |
XM_015284045.2 | XP_015139531.2 | 2 - 3 | 518 | 70 | ||
GenBank mRNA | U25176.1 | - | 2 - 3 | 536 | 70 | |
U67086.1 | - | 2 - 3 | 468 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
