Assay Details
| Gene Symbol: TPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204956.1 | NP_990287.1 | 8 - 9 | 1013 | 64 | |
| XM_015286577.2 | XP_015142063.2 | 8 - 9 | 1085 | 64 | ||
| GenBank mRNA | U26428.1 | - | 8 - 9 | 1013 | 64 | |
More Information
Set Membership: |
|

TPH1
Genomic Map