Assay Details
Gene Symbol: PTPN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204968.1 | NP_990299.1 | 2 - 3 | 248 | 73 | |
XM_025155426.1 | XP_025011194.1 | 2 - 3 | 73 | |||
GenBank mRNA | U38620.1 | - | 2 - 3 | 248 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
