Assay Details
Gene Symbol: DLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204973.2 | NP_990304.2 | 4 - 5 | 694 | 72 | |
XM_025148539.1 | XP_025004307.1 | 4 - 5 | 1617 | 72 | ||
XM_025148540.1 | XP_025004308.1 | 4 - 5 | 1617 | 72 | ||
XM_025148541.1 | XP_025004309.1 | 4 - 5 | 1617 | 72 | ||
GenBank mRNA | U26590.1 | - | 4 - 5 | 694 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
