Assay Details
| Gene Symbol: WNT9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204981.2 | NP_990312.2 | 3 - 4 | 665 | 79 | |
| GenBank mRNA | AF031168.1 | - | 3 - 4 | 665 | 79 | |
| AY753293.1 | - | 3 - 4 | 630 | 79 | ||
More Information
Set Membership: |
|

WNT9A
Genomic Map