Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204989.2 | NP_990320.2 | 2 - 3 | 181 | 78 | |
| GenBank mRNA | AF087432.1 | - | 2 - 3 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGG| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204989.2 | NP_990320.2 | 2 - 3 | 181 | 78 | |
| GenBank mRNA | AF087432.1 | - | 2 - 3 | 78 | ||
Set Membership: |
|