Assay Details
Gene Symbol: DEFB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201399.1 | NP_001188328.1 | 2 - 3 | 209 | 88 | |
NM_204992.2 | NP_990323.2 | 2 - 3 | 145 | 88 | ||
XM_015285091.2 | XP_015140577.1 | 2 - 3 | 162 | 88 | ||
GenBank mRNA | AF033336.1 | - | 1 - 2 | 88 | ||
BQ484799.1 | - | 2 - 3 | 88 | |||
CF250963.1 | - | 2 - 3 | 88 | |||
DQ677633.1 | - | 1 - 2 | 88 | |||
DQ858299.1 | - | 1 - 2 | 88 | |||
DQ858312.1 | - | 1 - 2 | 88 | |||
DQ858339.1 | - | 1 - 2 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
