Assay Details
| Gene Symbol: H2AFY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205007.1 | NP_990338.1 | 8 - 9 | 1079 | 84 | |
| XM_025154810.1 | XP_025010578.1 | 8 - 9 | 1115 | 84 | ||
| XM_025154811.1 | XP_025010579.1 | 8 - 9 | 1095 | 84 | ||
| XM_025154812.1 | XP_025010580.1 | 8 - 9 | 1106 | 84 | ||
| GenBank mRNA | AF058444.1 | - | 7 - 8 | 979 | 84 | |
| AF058445.1 | - | 8 - 9 | 1070 | 84 | ||
| AF058446.1 | - | 8 - 9 | 1079 | 84 | ||
| CR352888.1 | - | 5 - 6 | 674 | 84 | ||
| CR385822.1 | - | 4 - 5 | 583 | 84 | ||
More Information
Set Membership: |
|

H2AFY
Genomic Map