Assay Details
| Gene Symbol: XCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205046.1 | NP_990377.1 | 2 - 3 | 188 | 108 | |
| GenBank mRNA | AF006742.1 | - | 2 - 3 | 188 | 108 | |
| BX930561.1 | - | 2 - 3 | 210 | 108 | ||
| EU118364.1 | - | 2 - 3 | 178 | 108 | ||
More Information
Set Membership: |
|

XCL1
Genomic Map