Assay Details
| Gene Symbol: CD8BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205247.2 | NP_990578.2 | 3 - 4 | 890 | 116 | |
| GenBank mRNA | Y11475.1 | - | 490 | 116 | ||
| Z26484.1 | - | 890 | 116 | |||
More Information
Set Membership: |
|

CD8BP
Genomic Map