Assay Details
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205466.2 | NP_990797.2 | 3 - 4 | 355 | 85 | |
GenBank mRNA | J04614.1 | - | 3 - 4 | 349 | 85 | |
KP694235.1 | - | 3 - 4 | 362 | 85 | ||
KP694237.1 | - | 1 - 2 | 374 | 85 | ||
KP694238.1 | - | 2 - 3 | 259 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
