Assay Details
| Gene Symbol: POPDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206865.1 | NP_996747.1 | 2 - 3 | 934 | 72 | |
| XM_015294501.2 | XP_015149987.1 | 2 - 3 | 4576 | 72 | ||
| GenBank mRNA | AY388621.1 | - | 2 - 3 | 934 | 72 | |
| AY388622.1 | - | 2 - 3 | 934 | 72 | ||
| AY388623.1 | - | 2 - 3 | 934 | 72 | ||
| AY388624.1 | - | 2 - 3 | 934 | 72 | ||
| AY427076.1 | - | 2 - 3 | 934 | 72 | ||
More Information
Set Membership: |
|

POPDC2
Genomic Map