Assay Details
| Gene Symbol: CD247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206879.1 | NP_996762.1 | 3 - 4 | 265 | 62 | |
| XM_015295833.2 | XP_015151319.2 | 3 - 4 | 522 | 62 | ||
| GenBank mRNA | AJ002317.1 | - | 3 - 4 | 265 | 62 | |
More Information
Set Membership: |
|

CD247
Genomic Map