Assay Details
| Gene Symbol: CG-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206905.1 | NP_996788.1 | 1 - 2 | 88 | 64 | |
| XM_015276160.2 | XP_015131646.1 | 2 - 3 | 156 | 64 | ||
| GenBank mRNA | AY553270.1 | - | 1 - 2 | 88 | 64 | |
| BX930525.1 | - | 2 - 3 | 125 | 64 | ||
| BX933184.1 | - | 2 - 3 | 117 | 64 | ||
| BX935404.2 | - | 2 - 3 | 117 | 64 | ||
| BX935620.1 | - | 2 - 3 | 155 | 64 | ||
| M57240.1 | - | 2 - 3 | 123 | 64 | ||
More Information
Set Membership: |
|

CG-16
Genomic Map