Assay Details
Gene Symbol: SLC6A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213572.1 | NP_998737.1 | 3 - 4 | 842 | 63 | |
XM_015295699.2 | XP_015151185.1 | 4 - 5 | 3470 | 63 | ||
XM_015295700.2 | XP_015151186.1 | 4 - 5 | 3470 | 63 | ||
GenBank mRNA | AY573844.1 | - | 3 - 4 | 842 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
